The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136196.1 | NP_001129668.1 | 20 - 21 | 2490 | 76 |
GenBank mRNA | AF019039.1 | - | 20 - 21 | 2426 | 76 |