Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAGE1
Entrez Gene ID
285782
Gene Name
cancer antigen 1
Gene Aliases
CT3, CT95, CTAG3, bA69L16.7
Chromosome Location
Chr.6: 7326654 - 7389709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170692.1 NP_001164163.1 1 - 2 503 121
NM_001170693.1 NP_001164164.1 1 - 2 503 121
GenBank mRNA AK097760.1 - 1 - 2 202 121
BC026194.2 - 1 - 2 188 121
BC033545.1 - 1 - 2 188 121

View Full Product Details