The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286059.1 | NP_001272988.1 | 3 - 4 | 589 | 100 |
NM_001286060.1 | NP_001272989.1 | 3 - 4 | 424 | 100 | |
NM_001286061.1 | NP_001272990.1 | 3 - 4 | 322 | 100 | |
NM_014488.4 | NP_055303.2 | 4 - 5 | 525 | 100 | |
GenBank mRNA | AF498956.1 | - | 2 - 3 | 183 | 100 |
AK291376.1 | - | 3 - 4 | 584 | 100 | |
AK312323.1 | - | 3 - 4 | 330 | 100 | |
BC014213.2 | - | 4 - 5 | 444 | 100 | |
BP336371.1 | - | 3 - 4 | 322 | 100 | |
BX641138.1 | - | 3 - 4 | 373 | 100 | |
CR542106.1 | - | 2 - 3 | 183 | 100 | |
DB151524.1 | - | 3 - 4 | 425 | 100 |