The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015001.2 | NP_055816.2 | 12 - 13 | 10707 | 103 |
GenBank mRNA | AB023146.1 | - | 2 - 3 | 4502 | 103 |
AF356524.1 | - | 12 - 13 | 10707 | 103 | |
AL096858.1 | - | 9 - 10 | 9625 | 103 |