The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006231.3 | NP_006222.2 | 26 - 27 | 3486 | 90 |
XM_011534795.2 | XP_011533097.1 | 26 - 27 | 3481 | 90 | |
XM_011534797.2 | XP_011533099.1 | 18 - 19 | 2450 | 90 | |
XM_011534798.1 | XP_011533100.1 | 14 - 15 | 2002 | 90 | |
XM_011534799.2 | XP_011533101.1 | 26 - 27 | 3481 | 90 | |
XM_011534800.2 | XP_011533102.1 | 26 - 27 | 3481 | 90 | |
XM_011534802.2 | XP_011533104.1 | 2 - 3 | 303 | 90 | |
GenBank mRNA | L09561.1 | - | 26 - 27 | 3321 | 90 |
S60080.1 | - | 25 - 26 | 3203 | 90 | |
U49356.1 | - | 26 - 27 | 3274 | 90 |