Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNDP2
Entrez Gene ID
55748
Gene Name
CNDP dipeptidase 2 (metallopeptidase M20 family)
Gene Aliases
CN2, CPGL, HEL-S-13, HsT2298, PEPA
Chromosome Location
Chr.18: 74496265 - 74523454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168499.1 NP_001161971.1 4 - 5 876 134
NM_018235.2 NP_060705.2 7 - 8 1001 134
XM_005266728.2 XP_005266785.1 7 - 8 937 134
XM_006722503.2 XP_006722566.1 7 - 8 798 134
XM_011526071.2 XP_011524373.1 7 - 8 866 134
XM_011526072.2 XP_011524374.1 7 - 8 1821 134
XM_011526073.2 XP_011524375.1 6 - 7 1054 134
GenBank mRNA AF258592.1 - 4 - 5 604 134
AJ347717.1 - 6 - 7 765 134
AK001262.1 - 2 - 3 266 134
AK001692.1 - 7 - 8 904 134
AK024471.1 - 3 - 4 370 134
AK097155.1 - 7 - 8 1016 134
AK298309.1 - 7 - 8 620 134
AK300904.1 - 7 - 8 868 134
BC001375.2 - 7 - 8 874 134
BC003176.1 - 6 - 7 798 134
EU794600.1 - 6 - 7 798 134

View Full Product Details