The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000250.1 | NP_000241.1 | 7 - 8 | 1381 | 83 |
GenBank mRNA | BC130476.1 | - | 7 - 8 | 1255 | 83 |
J02694.1 | - | 7 - 8 | 1352 | 83 | |
M19507.1 | - | 7 - 8 | 1381 | 83 | |
S56200.1 | - | 7 - 8 | 1204 | 83 | |
X04876.1 | - | 7 - 8 | 1367 | 83 |