The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007231.4 | NP_009162.1 | 6 - 7 | 913 | 106 |
GenBank mRNA | AF151978.1 | - | 6 - 7 | 870 | 106 |
AK304243.1 | - | 6 - 7 | 781 | 106 | |
AK313390.1 | - | 6 - 7 | 915 | 106 | |
BC093710.1 | - | 6 - 7 | 804 | 106 | |
BC093712.1 | - | 6 - 7 | 804 | 106 |