The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292018.1 | NP_001278947.1 | 12 - 13 | 1476 | 96 |
NM_001292019.1 | NP_001278948.1 | 12 - 13 | 1042 | 96 | |
NM_004762.3 | NP_004753.1 | 12 - 13 | 1079 | 96 | |
NM_017456.3 | NP_059430.2 | 11 - 12 | 1076 | 96 | |
XM_011525475.2 | XP_011523777.1 | 11 - 12 | 938 | 96 | |
XM_011525476.2 | XP_011523778.1 | 11 - 12 | 938 | 96 | |
XM_011525477.1 | XP_011523779.1 | 10 - 11 | 857 | 96 | |
XM_011525478.1 | XP_011523780.1 | 11 - 12 | 1398 | 96 | |
XM_017025336.1 | XP_016880825.1 | 11 - 12 | 1146 | 96 | |
XM_017025337.1 | XP_016880826.1 | 11 - 12 | 1031 | 96 | |
XM_017025338.1 | XP_016880827.1 | 11 - 12 | 1462 | 96 | |
GenBank mRNA | AK293894.1 | - | 12 - 13 | 1476 | 96 |
AK296914.1 | - | 9 - 10 | 604 | 96 | |
AK307917.1 | - | 12 - 13 | 1008 | 96 | |
AK316277.1 | - | 12 - 13 | 1023 | 96 | |
BC038385.1 | - | 12 - 13 | 1000 | 96 | |
BC050452.1 | - | 12 - 13 | 989 | 96 | |
M85169.1 | - | 12 - 13 | 1029 | 96 |