Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIC1
Entrez Gene ID
57589
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Gene Aliases
CIP150, KIAA1432, bA207C16.1
Chromosome Location
Chr.9: 5629030 - 5778633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135920.2 NP_001129392.2 11 - 12 1443 93
NM_001206557.1 NP_001193486.1 11 - 12 1443 93
NM_020829.3 NP_065880.2 11 - 12 1443 93
XM_005251523.3 XP_005251580.1 11 - 12 1529 93
XM_011517967.2 XP_011516269.1 4 - 5 441 93
XM_017014934.1 XP_016870423.1 11 - 12 1099 93
XM_017014935.1 XP_016870424.1 9 - 10 2000 93
XM_017014936.1 XP_016870425.1 9 - 10 939 93
GenBank mRNA AB205401.1 - 10 - 11 1037 93
AK124394.1 - 8 - 9 1971 93
AL136875.1 - 11 - 12 1235 93
BC136616.1 - 10 - 11 1037 93
BC144296.1 - 10 - 11 1037 93

View Full Product Details