Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
Chromosome Location
Chr.9: 37602979 - 37746904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014907.2 NP_055722.2 10 - 11 1040 97
XM_011517803.2 XP_011516105.1 11 - 12 1442 97
XM_011517804.2 XP_011516106.1 10 - 11 1361 97
XM_011517805.2 XP_011516107.1 7 - 8 813 97
XM_011517806.2 XP_011516108.1 6 - 7 2049 97
XM_017014480.1 XP_016869969.1 10 - 11 1721 97
XM_017014481.1 XP_016869970.1 6 - 7 1416 97
XM_017014482.1 XP_016869971.1 7 - 8 1414 97
GenBank mRNA AB023184.1 - 10 - 11 1040 97
AK302739.1 - 7 - 8 1013 97
AK302854.1 - 6 - 7 626 97
BC114965.1 - 10 - 11 1019 97
BX648967.1 - 10 - 11 1590 97

View Full Product Details