Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
Chromosome Location
Chr.9: 37602979 - 37746904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014907.2 NP_055722.2 3 - 4 306 79
XM_011517803.2 XP_011516105.1 4 - 5 708 79
XM_011517804.2 XP_011516106.1 3 - 4 627 79
XM_017014480.1 XP_016869969.1 3 - 4 987 79
GenBank mRNA AB023184.1 - 3 - 4 306 79
BC114965.1 - 3 - 4 285 79
BX648967.1 - 3 - 4 856 79

View Full Product Details