Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
Chromosome Location
Chr.9: 37602979 - 37746904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014907.2 NP_055722.2 7 - 8 654 83
XM_011517803.2 XP_011516105.1 8 - 9 1056 83
XM_011517804.2 XP_011516106.1 7 - 8 975 83
XM_011517805.2 XP_011516107.1 4 - 5 427 83
XM_011517806.2 XP_011516108.1 3 - 4 1663 83
XM_017014480.1 XP_016869969.1 7 - 8 1335 83
XM_017014481.1 XP_016869970.1 3 - 4 1030 83
XM_017014482.1 XP_016869971.1 4 - 5 1028 83
GenBank mRNA AB023184.1 - 7 - 8 654 83
AK302739.1 - 4 - 5 627 83
AK302854.1 - 3 - 4 240 83
BC114965.1 - 7 - 8 633 83
BX648967.1 - 7 - 8 1204 83

View Full Product Details