The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282766.1 | NP_001269695.1 | 2 - 3 | 294 | 113 |
NM_022490.2 | NP_071935.1 | 2 - 3 | 294 | 113 | |
XM_005251547.2 | XP_005251604.1 | 1 - 2 | 539 | 113 | |
GenBank mRNA | AK023452.1 | - | 2 - 3 | 281 | 113 |
AK024032.1 | - | 2 - 3 | 270 | 113 | |
AK091294.1 | - | 1 - 2 | 655 | 113 | |
AK225050.1 | - | 2 - 3 | 267 | 113 | |
AK301347.1 | - | 2 - 3 | 284 | 113 | |
AK303167.1 | - | 2 - 3 | 281 | 113 | |
BC001337.1 | - | 2 - 3 | 261 | 113 | |
BC014331.1 | - | 2 - 3 | 295 | 113 | |
CR457313.1 | - | 2 - 3 | 181 | 113 |