Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR1E
Entrez Gene ID
64425
Gene Name
RNA polymerase I subunit E
Gene Aliases
PAF53, PRAF1
Chromosome Location
Chr.9: 37485935 - 37503697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282766.1 NP_001269695.1 7 - 8 749 93
NM_022490.2 NP_071935.1 6 - 7 655 93
XM_005251547.2 XP_005251604.1 5 - 6 900 93
GenBank mRNA AK023452.1 - 6 - 7 642 93
AK024032.1 - 6 - 7 631 93
AK091294.1 - 5 - 6 1016 93
AK225050.1 - 6 - 7 628 93
AK302345.1 - 5 - 6 632 93
AK303167.1 - 7 - 8 736 93
BC001337.1 - 6 - 7 622 93
BC014331.1 - 6 - 7 656 93
CR457313.1 - 6 - 7 542 93

View Full Product Details