Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ST18
- Entrez Gene ID
-
9705
- Gene Name
- ST18, C2H2C-type zinc finger
- Gene Aliases
-
NZF3, ZC2H2C3, ZC2HC10, ZNF387
- Chromosome Location
-
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC025662.1 |
- |
1 - 2 |
553 |
84 |
BC026316.1 |
- |
2 - 3 |
217 |
84 |
DA288225.1 |
- |
2 - 3 |
321 |
84 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI756594.1 |
- |
1 - 2 |
552 |
|
Chr. 8 - 52407658 - 52409875 [-] on Build GRCh38 |
84 |
GenBankmRNA |
HY132101.1 |
- |
2 - 3 |
248 |
|
Chr. 8 - 52407662 - 52409805 [-] on Build GRCh38 |
84 |
GenBankmRNA |
DA289724.1 |
- |
2 - 3 |
250 |
|
Chr. 8 - 52407678 - 52409808 [-] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101929341 |
LOC101929341 |
|
-208.1kb
|
Anti-sense |
Chr. - 52194362 - 52199552 on Build GRCh38 |
Downstream |
family with sequence similarity 150 member A |
FAM150A |
UNQ9433 |
124.2kb
|
Sense |
Chr. - 52534037 - 52565461 on Build GRCh38 |
View Full Product Details