The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000531.5 | NP_000522.3 | 4 - 5 | 603 | 118 |
XM_017029556.1 | XP_016885045.1 | 4 - 5 | 558 | 118 | |
GenBank mRNA | AK292757.1 | - | 4 - 5 | 558 | 118 |
BC074745.2 | - | 4 - 5 | 425 | 118 | |
BC107153.1 | - | 4 - 5 | 603 | 118 | |
BC107154.1 | - | 4 - 5 | 603 | 118 | |
BC114496.1 | - | 4 - 5 | 602 | 118 | |
K02100.1 | - | 4 - 5 | 524 | 118 |