The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022785.3 | NP_073622.2 | 29 - 30 | 4362 | 86 |
NM_198856.2 | NP_942153.1 | 27 - 28 | 4004 | 86 | |
XM_005261704.2 | XP_005261761.1 | 29 - 30 | 4302 | 86 | |
XM_005261705.3 | XP_005261762.1 | 28 - 29 | 4216 | 86 | |
XM_011530316.1 | XP_011528618.1 | 30 - 31 | 4589 | 86 | |
XM_011530317.2 | XP_011528619.1 | 28 - 29 | 4149 | 86 | |
XM_011530318.2 | XP_011528620.1 | 28 - 29 | 4245 | 86 | |
XM_011530319.2 | XP_011528621.1 | 27 - 28 | 3937 | 86 | |
XM_011530320.2 | XP_011528622.1 | 30 - 31 | 4401 | 86 | |
XM_011530321.2 | XP_011528623.1 | 27 - 28 | 3932 | 86 | |
XM_011530322.2 | XP_011528624.1 | 28 - 29 | 4060 | 86 | |
XM_011530323.2 | XP_011528625.1 | 25 - 26 | 3718 | 86 | |
XM_011530325.2 | XP_011528627.1 | 23 - 24 | 3463 | 86 | |
XM_011530327.2 | XP_011528629.1 | 20 - 21 | 3210 | 86 | |
XM_011530329.1 | XP_011528631.1 | 16 - 17 | 2682 | 86 | |
XM_017028910.1 | XP_016884399.1 | 30 - 31 | 4401 | 86 | |
XM_017028911.1 | XP_016884400.1 | 30 - 31 | 4401 | 86 | |
XM_017028912.1 | XP_016884401.1 | 26 - 27 | 4060 | 86 | |
GenBank mRNA | AB073862.1 | - | 9 - 10 | 1556 | 86 |
AK027241.1 | - | 9 - 10 | 1560 | 86 | |
AK225596.1 | - | 9 - 10 | 1561 | 86 | |
AK225605.1 | - | 9 - 10 | 1561 | 86 | |
AK292411.1 | - | 29 - 30 | 4302 | 86 | |
AK292498.1 | - | 27 - 28 | 3981 | 86 | |
AL355841.1 | - | 4 - 5 | 685 | 86 | |
BC039315.1 | - | 27 - 28 | 3857 | 86 |