The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005261704.2 | XP_005261761.1 | 31 - 32 | 4891 | 67 |
XM_005261705.3 | XP_005261762.1 | 30 - 31 | 4805 | 67 | |
XM_011530316.1 | XP_011528618.1 | 32 - 33 | 5178 | 67 | |
XM_011530317.2 | XP_011528619.1 | 30 - 31 | 4738 | 67 | |
XM_011530318.2 | XP_011528620.1 | 30 - 31 | 4834 | 67 | |
XM_011530319.2 | XP_011528621.1 | 29 - 30 | 4526 | 67 | |
XM_011530321.2 | XP_011528623.1 | 29 - 30 | 4521 | 67 | |
XM_011530322.2 | XP_011528624.1 | 30 - 31 | 4649 | 67 | |
XM_011530323.2 | XP_011528625.1 | 27 - 28 | 4307 | 67 | |
XM_011530325.2 | XP_011528627.1 | 25 - 26 | 4052 | 67 | |
XM_011530327.2 | XP_011528629.1 | 22 - 23 | 3799 | 67 | |
XM_011530329.1 | XP_011528631.1 | 18 - 19 | 3271 | 67 | |
XM_017028910.1 | XP_016884399.1 | 32 - 33 | 4990 | 67 | |
XM_017028912.1 | XP_016884401.1 | 28 - 29 | 4649 | 67 |