The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287742.1 | NP_001274671.1 | 8 - 9 | 1588 | 116 |
NM_001287743.1 | NP_001274672.1 | 7 - 8 | 1390 | 116 | |
NM_001287744.1 | NP_001274673.1 | 6 - 7 | 1217 | 116 | |
NM_001287745.1 | NP_001274674.1 | 7 - 8 | 1296 | 116 | |
NM_001287747.1 | NP_001274676.1 | 6 - 7 | 1253 | 116 | |
NM_001287748.1 | NP_001274677.1 | 6 - 7 | 915 | 116 | |
NM_001287749.1 | NP_001274678.1 | 6 - 7 | 1153 | 116 | |
NM_001287750.1 | NP_001274679.1 | 5 - 6 | 1155 | 116 | |
NM_001287751.1 | NP_001274680.1 | 5 - 6 | 1027 | 116 | |
NM_001287756.1 | NP_001274685.1 | 3 - 4 | 612 | 116 | |
NM_004462.4 | NP_004453.3 | 6 - 7 | 1099 | 116 | |
XM_017013217.1 | XP_016868706.1 | 8 - 9 | 1880 | 116 | |
GenBank mRNA | AK057726.1 | - | 6 - 7 | 1217 | 116 |
AK098682.1 | - | - | 677 | 116 | |
AK293545.1 | - | 6 - 7 | 1153 | 116 | |
AK296043.1 | - | 5 - 6 | 842 | 116 | |
AK297868.1 | - | 5 - 6 | 1098 | 116 | |
AK300059.1 | - | 5 - 6 | 917 | 116 | |
AK301617.1 | - | 3 - 4 | 473 | 116 | |
AK315993.1 | - | 7 - 8 | 1166 | 116 | |
AK316033.1 | - | 6 - 7 | 1120 | 116 | |
AK316182.1 | - | 5 - 6 | 1115 | 116 | |
AK316351.1 | - | 6 - 7 | 880 | 116 | |
AK316531.1 | - | 6 - 7 | 865 | 116 | |
AK316534.1 | - | 5 - 6 | 1027 | 116 | |
BC003573.1 | - | 6 - 7 | 946 | 116 | |
BC009251.2 | - | 6 - 7 | 946 | 116 | |
BC029641.1 | - | 6 - 7 | 909 | 116 | |
BC034440.1 | - | 6 - 7 | 909 | 116 | |
BT006704.1 | - | 6 - 7 | 877 | 116 | |
CR457033.1 | - | 6 - 7 | 877 | 116 | |
L06070.1 | - | 6 - 7 | 116 | ||
L06105.1 | - | 6 - 7 | 921 | 116 | |
S76822.1 | - | 6 - 7 | 942 | 116 | |
X69141.1 | - | 6 - 7 | 968 | 116 |