The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258033.1 | NP_001244962.1 | 8 - 9 | 723 | 66 |
NM_001258034.1 | NP_001244963.1 | 7 - 8 | 615 | 66 | |
NM_001258035.1 | NP_001244964.1 | 7 - 8 | 649 | 66 | |
NM_001258036.1 | NP_001244965.1 | 8 - 9 | 723 | 66 | |
NM_018119.3 | NP_060589.1 | 8 - 9 | 723 | 66 | |
NR_047581.1 | 8 - 9 | 673 | 66 | ||
XM_006721063.2 | XP_006721126.1 | 8 - 9 | 644 | 66 | |
XM_011545892.1 | XP_011544194.1 | 8 - 9 | 729 | 66 | |
XM_011545893.2 | XP_011544195.1 | 8 - 9 | 732 | 66 | |
XM_017023443.1 | XP_016878932.1 | 8 - 9 | 727 | 66 | |
GenBank mRNA | AB040885.1 | - | 8 - 9 | 643 | 66 |
AK001371.1 | - | 8 - 9 | 664 | 66 | |
AK023160.1 | - | 8 - 9 | 678 | 66 | |
AK023231.1 | - | 7 - 8 | 601 | 66 | |
AK026645.1 | - | 8 - 9 | 689 | 66 | |
AK222671.1 | - | 8 - 9 | 664 | 66 | |
AK222789.1 | - | 8 - 9 | 670 | 66 | |
AK293300.1 | - | 7 - 8 | 494 | 66 | |
AK296813.1 | - | 7 - 8 | 563 | 66 | |
AK300735.1 | - | 8 - 9 | 687 | 66 | |
AY092085.1 | - | 7 - 8 | 522 | 66 | |
BC000285.1 | - | 8 - 9 | 665 | 66 |