The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199867.1 | NP_001186796.1 | 1 - 2 | 385 | 74 |
NM_031417.3 | NP_113605.2 | 1 - 2 | 385 | 74 | |
GenBank mRNA | AB049127.1 | - | 1 - 2 | 351 | 74 |
AB058763.1 | - | 1 - 2 | 59 | 74 | |
AB088047.1 | - | 1 - 2 | 351 | 74 | |
AK075272.1 | - | 1 - 2 | 90 | 74 | |
AY057448.1 | - | 1 - 2 | 66 | 74 | |
AY120867.1 | - | 1 - 2 | 54 | 74 | |
DB100014.1 | - | 1 - 2 | 385 | 74 |