Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
Chromosome Location
Chr.19: 38647616 - 38730532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322033.1 NP_001308962.1 12 - 13 1576 65
NM_004924.5 NP_004915.2 12 - 13 1576 65
XM_005259281.4 XP_005259338.1 12 - 13 1576 65
XM_006723406.2 XP_006723469.1 12 - 13 1576 65
XM_017027331.1 XP_016882820.1 12 - 13 1576 65
GenBank mRNA AK304554.1 - 12 - 13 1505 65
AK309361.1 - 12 - 13 1505 65
AK309364.1 - 12 - 13 1505 65
BC005033.1 - 12 - 13 1459 65
BC015620.2 - 4 - 5 616 65
D89980.1 - 12 - 13 1453 65
GU987085.1 - 5 - 6 797 65
KU177881.1 - 12 - 13 1446 65

View Full Product Details