Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
Chromosome Location
Chr.19: 38647616 - 38730532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322033.1 NP_001308962.1 9 - 10 1045 107
NM_004924.5 NP_004915.2 9 - 10 1045 107
XM_005259281.4 XP_005259338.1 9 - 10 1045 107
XM_006723406.2 XP_006723469.1 9 - 10 1045 107
XM_017027331.1 XP_016882820.1 9 - 10 1045 107
GenBank mRNA AK299955.1 - 9 - 10 974 107
AK309361.1 - 9 - 10 974 107
AK309363.1 - 9 - 10 1012 107
AK309364.1 - 9 - 10 974 107
BC005033.1 - 9 - 10 928 107
BC015620.2 - 1 - 2 85 107
D89980.1 - 9 - 10 922 107
GU987085.1 - 2 - 3 266 107
KU177881.1 - 9 - 10 915 107

View Full Product Details