Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY4
Entrez Gene ID
57705
Gene Name
WDFY family member 4
Gene Aliases
C10orf64
Chromosome Location
Chr.10: 48684815 - 48982956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020945.1 NP_065996.1 41 - 42 7017 119
XM_005270004.3 XP_005270061.1 41 - 42 7132 119
XM_011539986.2 XP_011538288.2 41 - 42 7135 119
XM_011539987.2 XP_011538289.2 41 - 42 7138 119
XM_011539988.2 XP_011538290.1 42 - 43 7216 119
XM_011539990.2 XP_011538292.1 41 - 42 7163 119
XM_017016463.1 XP_016871952.1 42 - 43 7059 119
XM_017016464.1 XP_016871953.1 23 - 24 3446 119
GenBank mRNA AB046827.1 - 8 - 9 1200 119
AK074085.1 - 21 - 22 3075 119
AK074443.1 - 3 - 4 420 119
AL833669.1 - 1 - 2 146 119
BC047574.1 - 1 - 2 86 119

View Full Product Details