The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182520.2 | NP_872326.2 | 4 - 5 | 613 | 69 |
XM_011529907.2 | XP_011528209.1 | 3 - 4 | 1404 | 69 | |
XM_011529908.2 | XP_011528210.2 | 4 - 5 | 1125 | 69 | |
XM_011529912.2 | XP_011528214.1 | 2 - 3 | 1245 | 69 | |
XM_017028605.1 | XP_016884094.1 | 3 - 4 | 1419 | 69 | |
XM_017028611.1 | XP_016884100.1 | 3 - 4 | 1409 | 69 | |
XM_017028612.1 | XP_016884101.1 | 4 - 5 | 1109 | 69 | |
XM_017028615.1 | XP_016884104.1 | 4 - 5 | 1115 | 69 | |
GenBank mRNA | BU739625.1 | - | 2 - 3 | 69 |