The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135920.2 | NP_001129392.2 | 15 - 16 | 1879 | 100 |
NM_001206557.1 | NP_001193486.1 | 14 - 15 | 1768 | 100 | |
NM_020829.3 | NP_065880.2 | 15 - 16 | 1879 | 100 | |
XM_005251523.3 | XP_005251580.1 | 15 - 16 | 1965 | 100 | |
XM_011517967.2 | XP_011516269.1 | 8 - 9 | 877 | 100 | |
XM_011517968.2 | XP_011516270.1 | 3 - 4 | 212 | 100 | |
XM_017014934.1 | XP_016870423.1 | 15 - 16 | 1535 | 100 | |
XM_017014935.1 | XP_016870424.1 | 13 - 14 | 2436 | 100 | |
XM_017014936.1 | XP_016870425.1 | 13 - 14 | 1375 | 100 | |
GenBank mRNA | AB205401.1 | - | 14 - 15 | 1473 | 100 |
AK022642.1 | - | 1 - 2 | 99 | 100 | |
AK124394.1 | - | 11 - 12 | 2387 | 100 | |
AL136875.1 | - | 15 - 16 | 1671 | 100 | |
BC136616.1 | - | 14 - 15 | 1473 | 100 | |
BC144296.1 | - | 13 - 14 | 1362 | 100 |