Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
Chromosome Location
Chr.14: 34561409 - 34630160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021249.3 NP_067072.3 10 - 11 1068 141
NM_152233.2 NP_689419.2 11 - 12 963 141
XM_006720224.1 XP_006720287.1 10 - 11 850 141
GenBank mRNA AF109364.1 - 6 - 7 446 141
AF121856.1 - 11 - 12 954 141
AK021456.1 - 6 - 7 450 141
AK292250.1 - 11 - 12 946 141
AK296213.1 - 12 - 13 1091 141
AK308464.1 - 12 - 13 1091 141
BC001798.1 - 10 - 11 1068 141
BI870378.1 - 5 - 6 344 141
U83194.1 - 11 - 12 931 141

View Full Product Details