Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
Chromosome Location
Chr.14: 34561409 - 34630160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021249.3 NP_067072.3 5 - 6 672 140
NM_152233.2 NP_689419.2 6 - 7 567 140
XM_006720224.1 XP_006720287.1 5 - 6 454 140
GenBank mRNA AF121856.1 - 6 - 7 558 140
AK292250.1 - 6 - 7 550 140
AK296213.1 - 7 - 8 695 140
AK308464.1 - 7 - 8 695 140
BC001798.1 - 5 - 6 672 140
BG722853.1 - 6 - 7 594 141
U83194.1 - 6 - 7 535 140

View Full Product Details