Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 7 - 8 4616 108
NM_014866.1 NP_055681.1 8 - 9 4606 108
XM_005266134.2 XP_005266191.1 7 - 8 4469 108
XM_011519246.2 XP_011517548.1 8 - 9 4615 108
XM_011519247.1 XP_011517549.1 7 - 8 4472 108
XM_011519248.2 XP_011517550.1 7 - 8 4446 108
XM_011519249.1 XP_011517551.1 7 - 8 4469 108
XM_011519250.2 XP_011517552.1 7 - 8 4469 108
XM_011519251.1 XP_011517553.1 7 - 8 4469 108
XM_011519252.2 XP_011517554.1 7 - 8 4469 108
XM_011519253.1 XP_011517555.1 7 - 8 4469 108
XM_011519254.1 XP_011517556.1 7 - 8 4469 108
XM_011519255.2 XP_011517557.1 7 - 8 4469 108
XM_011519256.1 XP_011517558.1 7 - 8 4469 108
XM_011519257.1 XP_011517559.1 7 - 8 4469 108
XM_011519258.1 XP_011517560.1 7 - 8 4469 108
XM_011519259.1 XP_011517561.1 7 - 8 4469 108
XM_011519260.2 XP_011517562.1 7 - 8 4453 108
XM_011519261.1 XP_011517563.1 7 - 8 4469 108
XM_011519262.1 XP_011517564.1 8 - 9 3758 108
XM_011519263.2 XP_011517565.1 8 - 9 3758 108
XM_011519264.2 XP_011517566.1 7 - 8 4469 108
XM_017015363.1 XP_016870852.1 7 - 8 4469 108
XM_017015364.1 XP_016870853.1 7 - 8 4469 108
GenBank mRNA AB002308.4 - 8 - 9 4606 108
AB593115.1 - 7 - 8 4616 108
BC028183.1 - 3 - 4 473 108
BC125018.1 - 6 - 7 3178 108
BC125019.1 - 7 - 8 2467 108
DQ903855.1 - 6 - 7 3763 108

View Full Product Details