Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 10 - 11 4966 92
NM_014866.1 NP_055681.1 11 - 12 4956 92
XM_005266134.2 XP_005266191.1 10 - 11 4819 92
XM_011519246.2 XP_011517548.1 11 - 12 4965 92
XM_011519247.1 XP_011517549.1 10 - 11 4822 92
XM_011519248.2 XP_011517550.1 10 - 11 4796 92
XM_011519249.1 XP_011517551.1 10 - 11 4819 92
XM_011519250.2 XP_011517552.1 10 - 11 4819 92
XM_011519251.1 XP_011517553.1 10 - 11 4819 92
XM_011519252.2 XP_011517554.1 10 - 11 4819 92
XM_011519253.1 XP_011517555.1 10 - 11 4819 92
XM_011519254.1 XP_011517556.1 10 - 11 4819 92
XM_011519255.2 XP_011517557.1 10 - 11 4819 92
XM_011519256.1 XP_011517558.1 10 - 11 4819 92
XM_011519257.1 XP_011517559.1 10 - 11 4819 92
XM_011519258.1 XP_011517560.1 10 - 11 4819 92
XM_011519259.1 XP_011517561.1 10 - 11 4819 92
XM_011519260.2 XP_011517562.1 10 - 11 4803 92
XM_011519262.1 XP_011517564.1 11 - 12 4108 92
XM_011519263.2 XP_011517565.1 11 - 12 4108 92
XM_011519264.2 XP_011517566.1 10 - 11 4819 92
XM_017015363.1 XP_016870852.1 10 - 11 4819 92
GenBank mRNA AB002308.4 - 11 - 12 4956 92
AB593115.1 - 10 - 11 4966 92
BC028183.1 - 6 - 7 823 92
BC125018.1 - 9 - 10 3528 92
BC125019.1 - 10 - 11 2817 92
DQ903855.1 - 9 - 10 4113 92

View Full Product Details