Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELP
Entrez Gene ID
6403
Gene Name
selectin P
Gene Aliases
CD62, CD62P, GMP140, GRMP, LECAM3, PADGEM, PSEL
Chromosome Location
Chr.1: 169588849 - 169630139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003005.3 NP_002996.2 10 - 11 1766 122
XM_005245435.1 XP_005245492.1 10 - 11 1763 122
XM_005245436.3 XP_005245493.1 10 - 11 1763 122
XM_005245438.1 XP_005245495.1 10 - 11 1763 122
XM_005245439.1 XP_005245496.1 10 - 11 1763 122
XM_005245440.1 XP_005245497.1 9 - 10 1577 122
GenBank mRNA AK312884.1 - 10 - 11 1748 122
BC028067.2 - 9 - 10 1698 122
BC068533.1 - 10 - 11 1752 122
KU178327.1 - 9 - 10 1701 122
M25322.1 - 10 - 11 1739 122

View Full Product Details