Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELP
Entrez Gene ID
6403
Gene Name
selectin P
Gene Aliases
CD62, CD62P, GMP140, GRMP, LECAM3, PADGEM, PSEL
Chromosome Location
Chr.1: 169588849 - 169630139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003005.3 NP_002996.2 3 - 4 544 80
XM_005245435.1 XP_005245492.1 3 - 4 541 80
XM_005245436.3 XP_005245493.1 3 - 4 541 80
XM_005245438.1 XP_005245495.1 3 - 4 541 80
XM_005245439.1 XP_005245496.1 3 - 4 541 80
XM_005245440.1 XP_005245497.1 3 - 4 541 80
GenBank mRNA AK312884.1 - 3 - 4 526 80
AY367063.1 - 1 - 2 133 80
BC028067.2 - 2 - 3 476 80
BC068533.1 - 3 - 4 530 80
KU178327.1 - 2 - 3 479 80
M25322.1 - 3 - 4 517 80

View Full Product Details