The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020729.2 | NP_065780.2 | 15 - 16 | 1924 | 152 |
GenBank mRNA | BC047368.1 | - | 15 - 16 | 1864 | 152 |