The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007357.2 | NP_031383.1 | 14 - 15 | 1804 | 125 |
GenBank mRNA | AK295251.1 | - | 14 - 15 | 1811 | 125 |
AK302288.1 | - | 6 - 7 | 777 | 125 | |
AK302791.1 | - | 15 - 16 | 1988 | 125 | |
AK312678.1 | - | 14 - 15 | 1720 | 125 | |
BC014960.1 | - | 14 - 15 | 1767 | 125 | |
Z34975.1 | - | 14 - 15 | 1748 | 125 |