The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020368.2 | NP_065101.1 | 1 - 1 | 1892 | 122 |
GenBank mRNA | AF271212.1 | - | 1 - 1 | 1854 | 122 |
AK026909.1 | - | 1 - 1 | 1841 | 122 | |
AL136590.1 | - | 1 - 1 | 1827 | 122 |