The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170331.1 | NP_001163802.1 | 3 - 4 | 1200 | 90 |
NM_198511.2 | NP_940913.1 | 3 - 4 | 1200 | 90 | |
XM_011543904.2 | XP_011542206.1 | 3 - 4 | 685 | 90 | |
GenBank mRNA | AK124915.1 | - | 3 - 4 | 1200 | 90 |
BC093667.1 | - | 3 - 4 | 992 | 90 | |
BC093669.1 | - | 3 - 4 | 992 | 90 | |
BC143528.1 | - | 3 - 4 | 992 | 90 |