The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170331.1 | NP_001163802.1 | 4 - 5 | 1404 | 77 |
XM_011543904.2 | XP_011542206.1 | 4 - 5 | 889 | 77 | |
GenBank mRNA | BC143528.1 | - | 4 - 5 | 1196 | 77 |