The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004092.3 | NP_004083.3 | 6 - 7 | 805 | 111 |
GenBank mRNA | BC008906.2 | - | 6 - 7 | 748 | 111 |
BC013264.1 | - | 6 - 7 | 740 | 111 | |
BQ130816.1 | - | 4 - 5 | 440 | 111 | |
BT007123.1 | - | 6 - 7 | 734 | 111 | |
D13900.1 | - | 6 - 7 | 755 | 111 |