The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153646.3 | NP_705932.2 | 9 - 10 | 757 | 94 |
NM_153647.3 | NP_705933.2 | 10 - 11 | 830 | 94 | |
NM_153648.3 | NP_705934.1 | 9 - 10 | 659 | 94 | |
XM_005267342.2 | XP_005267399.1 | 8 - 9 | 426 | 94 | |
XM_011536436.2 | XP_011534738.1 | 10 - 11 | 980 | 94 | |
XM_011536437.2 | XP_011534739.1 | 10 - 11 | 980 | 94 | |
XM_011536438.2 | XP_011534740.1 | 10 - 11 | 980 | 94 | |
XM_011536439.2 | XP_011534741.1 | 10 - 11 | 980 | 94 | |
GenBank mRNA | AF520704.1 | - | 9 - 10 | 707 | 94 |
AF520705.1 | - | 9 - 10 | 707 | 94 | |
AF520706.1 | - | 9 - 10 | 659 | 94 | |
AK096171.1 | - | 8 - 9 | 432 | 94 | |
AK295059.1 | - | 10 - 11 | 830 | 94 | |
AK295234.1 | - | 3 - 4 | 187 | 94 | |
BC069653.1 | - | 9 - 10 | 707 | 94 |