The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080414.3 | NP_001073883.2 | 6 - 7 | 637 | 58 |
XM_005267691.4 | XP_005267748.1 | 6 - 7 | 1291 | 58 | |
XM_011536796.2 | XP_011535098.1 | 6 - 7 | 591 | 58 | |
XM_017021335.1 | XP_016876824.1 | 6 - 7 | 1291 | 58 | |
XM_017021337.1 | XP_016876826.1 | 6 - 7 | 1291 | 58 | |
GenBank mRNA | AK302839.1 | - | 5 - 6 | 417 | 58 |
DC402942.1 | - | 5 - 6 | 417 | 58 | |
EG328265.1 | - | 1 - 2 | 30 | 58 |