Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 15 - 16 1922 117
NM_020784.2 NP_065835.2 15 - 16 1937 117
GenBank mRNA AB037765.2 - 15 - 16 1849 117
AK022563.1 - 1 - 2 123 117
AK097748.1 - 16 - 17 1826 117
BC137081.1 - 15 - 16 1817 117
BC137082.1 - 15 - 16 1817 117
BC142650.1 - 15 - 16 1860 117
BC144469.1 - 15 - 16 1802 117
BC150205.1 - 15 - 16 1858 117
BC152428.1 - 15 - 16 1849 117

View Full Product Details