Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 4 - 5 687 150
NM_020784.2 NP_065835.2 4 - 5 687 150
XM_017021505.1 XP_016876994.1 4 - 5 667 150
GenBank mRNA AB037765.2 - 4 - 5 599 150
AK097748.1 - 4 - 5 474 150
BC137081.1 - 4 - 5 567 150
BC137082.1 - 4 - 5 567 150
BC142650.1 - 4 - 5 610 150
BC144469.1 - 4 - 5 567 150
BC150205.1 - 4 - 5 608 150
BC152428.1 - 4 - 5 599 150

View Full Product Details