Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
Chromosome Location
Chr.11: 65961689 - 65980136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005146.4 NP_005137.1 2 - 3 465 65
XM_011545344.1 XP_011543646.1 2 - 3 494 65
XM_011545345.2 XP_011543647.1 2 - 3 216 65
GenBank mRNA AB006198.1 - 2 - 3 411 65
AF353625.1 - 2 - 3 406 65
AK223016.1 - 2 - 3 426 65
AK293200.1 - 3 - 4 332 65
AK297589.1 - 2 - 3 426 65
BC001058.2 - 2 - 3 414 65
BT006637.1 - 2 - 3 373 65
BX648500.1 - 2 - 3 215 65
Y14314.1 - 2 - 3 247 65

View Full Product Details