Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
Chromosome Location
Chr.11: 65961689 - 65980136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005146.4 NP_005137.1 5 - 6 751 65
XM_011545344.1 XP_011543646.1 5 - 6 780 65
XM_011545345.2 XP_011543647.1 5 - 6 543 65
GenBank mRNA AB006198.1 - 5 - 6 697 65
AF353625.1 - 5 - 6 692 65
AK293200.1 - 6 - 7 618 65
AK297589.1 - 5 - 6 753 65
BC001058.2 - 5 - 6 700 65
BT006637.1 - 5 - 6 659 65
BX648500.1 - 5 - 6 501 65
Y14314.1 - 5 - 6 533 65

View Full Product Details