Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
Chromosome Location
Chr.11: 65961689 - 65980136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005146.4 NP_005137.1 9 - 10 1276 63
XM_011545344.1 XP_011543646.1 9 - 10 1305 63
XM_011545345.2 XP_011543647.1 9 - 10 1068 63
GenBank mRNA AB006198.1 - 9 - 10 1222 63
AF353625.1 - 9 - 10 1217 63
AK223016.1 - 9 - 10 1237 63
AK293200.1 - 10 - 11 1143 63
AK297589.1 - 8 - 9 1409 63
BC001058.2 - 9 - 10 1225 63
BC018607.1 - 1 - 2 108 63
BT006637.1 - 9 - 10 1184 63
BX648500.1 - 9 - 10 1026 63
Y14314.1 - 9 - 10 1058 63

View Full Product Details