Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTAGE11P
Entrez Gene ID
647288
Gene Name
CTAGE family member 11, pseudogene
Gene Aliases
-
Chromosome Location
Chr.13: 75237753 - 75240381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027466.1 1 - 1 2380 120
GenBank mRNA AL133018.1 - 1 - 1 2380 120

View Full Product Details