The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033348.1 | 1 - 2 | 320 | 90 | |
XM_011545439.2 | XP_011543741.1 | 1 - 2 | 282 | 90 | |
GenBank mRNA | BC027937.1 | - | 1 - 2 | 200 | 90 |