The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130777.2 | NP_570133.1 | 4 - 5 | 92 | |
XM_011530836.2 | XP_011529138.1 | 4 - 5 | 92 | ||
GenBank mRNA | AI740470.1 | - | 1 - 2 | 127 | 92 |
AJ318880.1 | - | 4 - 5 | 92 | ||
BC066311.1 | - | 4 - 5 | 92 |