Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAGE3
Entrez Gene ID
139793
Gene Name
PAGE family member 3
Gene Aliases
CT16.6, GAGED1, PAGE-3
Chromosome Location
Chr.X: 55257833 - 55264916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171252.1 NP_001164723.1 2 - 3 411 134
GenBank mRNA BC144449.1 - 2 - 3 296 134

View Full Product Details